Dimedone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethylcyclohexane-1,3-dione

A10140-14 A10140-22 A10140-36
CAAAA10140-14EA 57.35 CAD
CAAAA10140-14 CAAAA10140-22 CAAAA10140-36
Dimedone 98%
Dimedone

Reagent used in synthesis of fused 3-methylfurans


Purity: 98% CAS No.: 126-81-8 Molecular Formula: C8H12O2 Formula Weight: 140.18 Melting Point: 146-150° Beilstein Reference: 471489 Merck Reference: 14,3242 MDL No.: MFCD00001588

Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00001588
CAS Number: 126-81-8
EINECS: 204-804-4
Merck Index: 14,3242

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR