1,4-Dithioerythritol 99%

Supplier: Thermo Scientific Chemicals
Warning

A10138-06 A10138-14
CAAAA10138-06EA 211.72 CAD
CAAAA10138-06 CAAAA10138-14
1,4-Dithioerythritol 99%
1,4-Dithioerythritol

Reagent for maintaining thiols in reduced state.


Purity: 99% CAS No.: 6892-68-8 Molecular Formula: C4H10O2S2 Formula Weight: 154.25 Storage Temperature: 2°C to 8°C Odor: Stench Beilstein Reference: 1719756 UN No.: 3335 MDL No.: MFCD00063750

Formula: C₄H₁₀O₂S₂
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 82…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063750
CAS Number: 6892-68-8
EINECS: 229-998-8
UN: 3335
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR