5-Bromo-3-methoxysalicylaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10137-06
A10137-14
A10137-22
CAAAA10137-06EA
74.26
CAD
CAAAA10137-06
CAAAA10137-14
CAAAA10137-22
5-Bromo-3-methoxysalicylaldehyde 98%
5-Bromo-3-methoxysalicylaldehyde
MDL: MFCD00016593
Beilstein Registry No.: 2328601
Purity: 98%
CAS No.: 5034-74-2
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
Melting Point: 127-131°
Beilstein Reference: 2328601
Formula:
C₈H₇BrO₃ Boiling Pt: 314 °C (1013 hPa) Melting Pt: 127…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016593 CAS Number: 5034-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...