DL-α-Phenylalanine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-DL-Phe-OH, DL-Phenylalanine, 3-Phenyl-DL-alanine

A10132-14 A10132-22 A10132-36
CAAAA10132-14EA 39.66 CAD
CAAAA10132-14 CAAAA10132-22 CAAAA10132-36
DL-α-Phenylalanine 99%
DL-α-Phenylalanine

MDL: MFCD00064225 Beilstein Registry No.: 1910407


Purity: 99% CAS No.: 150-30-1 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: ca 266° dec. Beilstein Reference: 1910407 Merck Reference: 14,7271 MDL No.: MFCD00064225

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 313
Melting Pt: 173 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064225
CAS Number: 150-30-1
EINECS: 205-756-7
Merck Index: 14,7271

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR