1,2,4-Trichlorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10130-0E
A10130-36
CAAAA10130-0EEA
205.4
CAD
CAAAA10130-0E
CAAAA10130-36
1,2,4-Trichlorobenzene 99%
1,2,4-Trichlorobenzene
MDL: MFCD00000547
Miscible with ether, benzene, CS2
Purity: 99%
CAS No.: 120-82-1
Molecular Formula: C6H3Cl3
Formula Weight: 181.45
Flash Point: 110°(230°F)
Refractive Index: 1.572
Melting Point: 17°
Boiling Point: 213°
Solubility: Miscible with ether, benzene, CS2
Merck Reference: 14,9631
UN No.: 2321
MDL No.: MFCD00000547
Formula:
C₆H₃Cl₃ MW: 181.45 g/mol Boiling Pt: 214.4 Melting Pt: 17 Density: 1.454 Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000547 CAS Number: 120-82-1 EINECS: 204-428-0 UN: 2321 ADR: 6.1 III Merck Index: 14,9631 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...