4-Fluorobenzyl mercaptan 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Fluorotoluene-α-thiol, 4-Fluorobenzylmercaptan

A10126-03 A10126-06
CAAAA10126-03EA 60.83 CAD
CAAAA10126-03 CAAAA10126-06
4-Fluorobenzyl mercaptan 96%
4-Fluorobenzyl mercaptan

MDL: MFCD00040724 Beilstein Registry No.: 2242520


Purity: 96% CAS No.: 15894-04-9 Molecular Formula: C7H7FS Formula Weight: 142.20 Flash Point: 76°(168°F) Refractive Index: 1.5430 Boiling Point: 72-74°/15mm Beilstein Reference: 2242520 UN No.: 2810 MDL No.: MFCD00040724

Formula: C₇H₇FS
MW: 142.2
Boiling Pt: 37
Density: 1.157
Flash Pt: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040724
CAS Number: 15894-04-9
EINECS: 240-031-9
UN: 3334
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR