4-(Bromomethyl)benzenesulfonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Bromomethyl)benzene-1-sulfonyl chloride, 4-(Bromomethyl)benzene-1-sulphonyl chloride

A10120-06 A10120-14
CAAAA10120-06EA 155.31 CAD
CAAAA10120-06 CAAAA10120-14
4-(Bromomethyl)benzenesulfonyl chloride 95%
4-(Bromomethyl)benzenesulphonyl chloride

MDL: MFCD00156129 Beilstein Registry No.: 2364328

Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00156129
CAS Number: 66176-39-4
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR