2,4-Dichlorobenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10113-06
A10113-14
CAAAAA10113-22EA
961.67
CAD
CAAAAA10113-22
CAAAA10113-06
CAAAA10113-14
2,4-Dichlorobenzonitrile 98%
2,4-Dichlorobenzonitrile
MDL: MFCD00016373
Beilstein Registry No.: 637738
Purity: 98%
CAS No.: 6574-98-7
Molecular Formula: C7H3Cl2N
Formula Weight: 172.01
Melting Point: 57-61°
Beilstein Reference: 637738
UN No.: 3439
MDL No.: MFCD00016373
Formula:
C₇H₃Cl₂N Boiling Pt: 261 °C (1013 hPa) Melting Pt: 57…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016373 CAS Number: 6574-98-7 EINECS: 229-493-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...