Methyl-4-chlorobenzoate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Chlorobenzoic acid emthyl ester, Methyl 4-chlorobenzoate
A10109-14
A10109-22
CAAAA10109-14EA
54.19
CAD
CAAAA10109-14
CAAAA10109-22
Methyl-4-chlorobenzoate 99%
Methyl 4-chlorobenzoate
MDL: MFCD00000621
Beilstein Registry No.: 1564310
Purity: 99%
CAS No.: 1126-46-1
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Flash Point: 106°(222°F)
Melting Point: 41-45°
Boiling Point: 136-137°/10mm
Beilstein Reference: 1564310
Merck Reference: 14,2125
MDL No.: MFCD00000621
Formula:
C₈H₇ClO₂ Boiling Pt: 136…137 °C/10 mm Melting Pt: 41…45 °C Density: 1.382 Flash Pt: 106 °C(222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000621 CAS Number: 1126-46-1 EINECS: 214-420-9 Merck Index: 14,2125 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...