2-Chloropyrazine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chloropyrazine

A10108-06 A10108-14 A10108-22
CAAAA10108-06EA 79.95 CAD
CAAAA10108-06 CAAAA10108-14 CAAAA10108-22
2-Chloropyrazine 98%
2-Chloropyrazine

MDL: MFCD00006124 Beilstein Registry No.: 107019


Purity: 98% CAS No.: 14508-49-7 Molecular Formula: C4H3ClN2 Formula Weight: 114.54 Flash Point: 57°(134°F) Refractive Index: 1.5370 Boiling Point: 153-154° Beilstein Reference: 107019 UN No.: 1993 MDL No.: MFCD00006124

Formula: C₄H₃ClN₂
MW: 114.53 g/mol
Boiling Pt: 153…154 °C
Density: 1.284
Flash Pt: 57 °C (134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006124
CAS Number: 14508-49-7
EINECS: 238-517-0
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR