2,6-Dichlorophenolindophenol sodium salt hydrate

Supplier: Thermo Scientific Chemicals

A10107-06 A10107-14 A10107-22
CAAAA10107-06EA 111.23 CAD
CAAAA10107-06 CAAAA10107-14 CAAAA10107-22
2,6-Dichlorophenolindophenol sodium salt hydrate
2,6-Dichlorophenolindophenol sodium salt hydrate

Suitable for ascorbic acid determination


2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase.


It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.

Formula: C₁₂H₆Cl₂NNaO₂·H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150014
CAS Number: 620-45-1
EINECS: 210-640-4
Merck Index: 14,3068

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR