3-Chloro-4-methoxybenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A10106-06 A10106-14
CAAAA10106-06EA 108.55 CAD
CAAAA10106-06 CAAAA10106-14 CAAAA10106-22
3-Chloro-4-methoxybenzoic acid 98+%
3-Chloro-4-methoxybenzoic acid

MDL: MFCD00016512 Beilstein Registry No.: 2087584


Purity: 98+% CAS No.: 37908-96-6 Molecular Formula: C8H7ClO3 Formula Weight: 186.59 Melting Point: 215-219° Beilstein Reference: 2087584

Formula: C₈H₇ClO₃
Melting Pt: 215…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00016512
CAS Number: 37908-96-6
EINECS: 253-708-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR