2-Bromo-5-chlorobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A10103-03 A10103-06 A10103-14
CAAAA10103-03EA 21.49 CAD
CAAAA10103-03 CAAAA10103-06 CAAAA10103-14
2-Bromo-5-chlorobenzoic acid 98+%
2-Bromo-5-chlorobenzoic acid

MDL: MFCD00013982 Beilstein Registry No.: 2442261


Purity: 98+% CAS No.: 21739-93-5 Molecular Formula: C7H4BrClO2 Formula Weight: 235.46 Odor: Odorless Melting Point: 152-156° Beilstein Reference: 2442261 UN No.: 2811 MDL No.: MFCD00013982

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00013982
CAS Number: 21739-93-5
EINECS: 244-559-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR