Diphosphorus pentoxide ≥99.99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
89966-09
89966-22
89966-36
CAAA89966-09EA
39.82
CAD
CAAA89966-09
CAAA89966-22
CAAA89966-36
Diphosphorus pentoxide ≥99.99%
di-Phosphorus pentoxide
MDL: MFCD00011440
Fieser: 1,871 7,291 10,319 15,267 18,293 20,309
Purity: 99.99%
CAS No.: 1314-56-3
Molecular Formula: P2O5
Formula Weight: 141.94
Physical Form: Powder
Density: 2.39
Melting Point: 340°
Boiling Point: 360° subl.
Merck Reference: 14,7355
UN No.: 1807
MDL No.: MFCD00011440
Formula:
P₂O₅ Boiling Pt: 360 °C (subl.) Melting Pt: 340 °C Density: 2.39 Storage Temperature: Ambient |
MDL Number:
MFCD00011440 CAS Number: 1314-56-3 EINECS: 215-236-1 UN: 1807 ADR: 8,II Merck Index: 14,7355 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...