Diboron trioxide ≥99.98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
89964-22
89964-36
CAAA89964-22EA
142.2
CAD
CAAA89964-22
CAAA89964-36
Diboron trioxide ≥99.98% (metals basis)
di-Boron trioxide
MDL: MFCD00011315
Purity: 99.98%
CAS No.: 1303-86-2
Molecular Formula: B2O3
Formula Weight: 69.62
Physical Form: Powder
Density: 2.46
Appearance: White
Odor: Odorless
Melting Point: ca 450°
Boiling Point: 1860°
Merck Reference: 14,1337
MDL No.: MFCD00011315
Formula:
B₂O₃ Boiling Pt: 1860 °C Melting Pt: ∼450 °C Density: 2.46 Storage Temperature: Ambient |
MDL Number:
MFCD00011315 CAS Number: 1303-86-2 EINECS: 215-125-8 REACH: 01-2119486655-24 Merck Index: 14,1337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...