Erbium trichloride, anhydrous ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
89917-09
89917-18
CAAAA89917-30EA
7670.3
CAD
CAAAA89917-30
CAAA89917-09
CAAA89917-18
Erbium trichloride, anhydrous ≥99.9% (metals basis)
Erbium trichloride
Anhydrous
Purity: 99.9%
CAS No.: 10138-41-7
Molecular Formula: ErCl3
Formula Weight: 273.62
Physical Form: Beads
Sensitivity: Hygroscopic
Boiling Point: 1500°
MDL No.: MFCD00010989
Formula:
Cl₃Er Boiling Pt: 1500 °C Melting Pt: 774 °C Density: 4.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010989 CAS Number: 10138-41-7 EINECS: 233-385-0 |
Specification Test Results
Form | Beads |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...