Samarium trichloride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

89916-09 89916-18
CAAA89916-09EA 214.88 CAD
CAAA89916-09 CAAA89916-18
Samarium trichloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Samarium trichloride

MDL: MFCD00011235


Purity: 99.9% CAS No.: 10361-82-7 Molecular Formula: SmCl3 Formula Weight: 256.71 Physical Form: Chunks/Powder Density: 4.46 Melting Point: 678° Boiling Point: dec. Merck Reference: 14,8347 MDL No.: MFCD00011235

Formula: Cl₃Sm
Boiling Pt: Dec.
Melting Pt: 678 °C
Density: 4.46
Storage Temperature: Ambient
MDL Number: MFCD00011235
CAS Number: 10361-82-7
EINECS: 233-797-0
Merck Index: 14,8347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR