Aluminium chloride, anhydrous ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum chloride
89914-A1
89914-D9
CAAA89914-A1EA
105.7
CAD
CAAA89914-A1
CAAA89914-D9
Aluminium chloride, anhydrous ≥95%
Aluminium chloride
MDL: MFCD00003422
Fieser: 1,24 12,26 13,15 14,21 15,10 16,10 17,15 18,18 19,11 20,13
Purity: 95+%
CAS No.: 7446-70-0
Molecular Formula: AlCl3
Formula Weight: 133.34
Physical Form: Powder
Density: 2.44
Appearance: Yellow to gray
Odor: Pungent
pH: 2,4
Melting Point: 190° subl.
Merck Reference: 14,337
UN No.: 1726
MDL No.: MFCD00003422
Formula:
AlCl₃ MW: 133.34 g/mol Boiling Pt: 180 °C (1013 hPa) Melting Pt: 190 °C (subl.) Density: 2.44 Storage Temperature: Ambient |
MDL Number:
MFCD00003422 CAS Number: 7446-70-0 EINECS: 231-208-1 UN: 1726 ADR: 8,II Merck Index: 14,337 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...