Strontium sulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

89895-09 89895-18
CAAA89895-09EA 195.92 CAD
CAAA89895-09 CAAA89895-18
Strontium sulfide ≥99.9% (metals basis)
Strontium sulphide

MDL: MFCD00049560 Slightly soluble in water. Decomposes in acids


Purity: 99.9% CAS No.: 1314-96-1 Molecular Formula: SrS Formula Weight: 119.68 Physical Form: Powder Density: 3.70 Appearance: Grey Odor: Like rotten eggs Melting Point: 2000° Solubility: Slightly soluble in water. Decomposes in acids Merck Reference: 14,8849 MDL No.: MFCD00049560

Formula: SSr
Melting Pt: >2000 °C
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00049560
CAS Number: 1314-96-1
EINECS: 215-249-2
UN: 3134
ADR: 4.3,III
Merck Index: 14,8849

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR