Ytterbium, AAS Standard Solution, Specpure®, Yb 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
89890-AE
89890-AP
CAAA89890-AEEA
65.41
CAD
CAAA89890-AE
CAAA89890-AP
Ytterbium, AAS Standard Solution, Specpure®, Yb 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Ytterbium Standards
MDL: MFCD00011286
Molecular Formula: Yb2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011286
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...