Calcium chloride, anhydrous ≥96.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

89866-18 89866-30
CAAA89866-18EA 108.23 CAD
CAAA89866-18 CAAA89866-30
Calcium chloride, anhydrous ≥96.0% ACS
Calcium chloride

Typical ionic halide, often used as a dessicant


Purity: 96.0% min CAS No.: 10043-52-4 Molecular Formula: CaCl2 Formula Weight: 110.99 Physical Form: Powder, Packaged under argon Sensitivity: Hygroscopic Boiling Point: 1600° Merck Reference: 14,1659 MDL No.: MFCD00010903


Typical ionic halide, often used as a dessicant

Formula: CaCl₂
Boiling Pt: >1600 °C
Melting Pt: 782 °C
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,1659

Order Now

Specification Test Results

Titratable base 0.006meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR