Lanthanum(III) sulfate, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Dilanthanum trisulfate, Dilanthanum trisulphate
89760-18
89760-30
CAAA89760-18EA
180.12
CAD
CAAA89760-18
CAAA89760-30
Lanthanum(III) sulfate, anhydrous ≥99.9% (REO, rare earth oxide basis)
Lanthanum (III) sulphate
MDL: MFCD00011070
Purity: 99.9%
CAS No.: 10099-60-2
Molecular Formula: La2(SO4)3
Formula Weight: 566.00
Physical Form: Powder
Density: 3.60
Appearance: White
Odor: Odorless
Melting Point: 1150° dec.
Merck Reference: 14,5363
MDL No.: MFCD00011070
Formula:
La₂O₁₂S₃ MW: 566 Melting Pt: 1150 °C (dec.) Density: 3.60 Storage Temperature: Ambient |
MDL Number:
MFCD00011070 CAS Number: 10099-60-2 EINECS: 233-239-6 Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...