Silicon dioxide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

89709-30 89709-A1 89709-A7
CAAA89709-30EA 103.17 CAD
CAAA89709-30 CAAA89709-A1 CAAA89709-A7
Silicon dioxide ≥99.5% (metals basis)
Silicium dioxide

MDL: MFCD00011232


Purity: 99.5% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Fused Amorphous Powder, S.A. 5m2/g Density: 2.2 pH: 6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1719
Density: 2.2
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,08493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR