Niobium(IV) oxide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Niobium dioxide

89692-09 89692-18
CAAA89692-09EA 173.8 CAD
CAAA89692-09 CAAA89692-18
Niobium(IV) oxide ≥99% (metals basis)
Niobium (IV) oxide

MDL: MFCD00016242


Purity: 99+% CAS No.: 12034-59-2 Molecular Formula: NbO2 Formula Weight: 124.91 Physical Form: Powder Density: 5.9 MDL No.: MFCD00016242

Formula: NbO₂
MW: 124.91 g/mol
Melting Pt: 1915
Density: 5.9
Storage Temperature: Ambient
MDL Number: MFCD00016242
CAS Number: 12034-59-2
EINECS: 234-809-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR