Lithium aluminum hydride ≥97%, pellets
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminium lithium hydride, Lithium tetrahydridoaluminate
89643-14
89643-22
89643-36
CAAA89643-14EA
116.29
CAD
CAAA89643-14
CAAA89643-22
CAAA89643-36
Lithium aluminum hydride ≥97%, pellets
Lithium aluminum hydride
MDL: MFCD00011075
Fieser: 1,581 14,190 15,184 17,162 18,207 19,191 20,213 21,247
Purity: 97%
CAS No.: 16853-85-3
Molecular Formula: LiAlH4
Formula Weight: 37.95
Physical Form: Pellets
Density: 0.917
Appearance: Whote to grey
Odor: Odorless
Melting Point:
Formula:
H₄AlLi MW: 37.95 g/mol Melting Pt: >125 °C (dec.) Density: 0.917 Storage Temperature: Ambient |
MDL Number:
MFCD00011075 CAS Number: 16853-85-3 EINECS: 240-877-9 UN: 1410 ADR: 4.3,I Merck Index: 14,349 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...