Chloroform-[D₁] (99.8% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

89541-18 89541-30
CAAA89541-18EA 145.68 CAD
CAAA89541-18 CAAA89541-30
Chloroform-[D₁] (99.8% D) (Isotopic) for NMR spectroscopy
Chloroform-[D1]

NMR solvent


Purity: 99.8% CAS No.: 865-49-6 Molecular Formula: CDCl3 Formula Weight: 120.38 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.50 Refractive Index: 1.4445 Sensitivity: Moisture and Light Sensitive Appearance: Colorless Melting Point: -64° Boiling Point: 62° UN No.: 1888 MDL No.: MFCD00000827


NMR solvent

Formula: CDCl₃
Boiling Pt: 62 °C
Melting Pt: –64 °C
Density: 1.50
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR