Bismuth(III) chloride, anhydrous ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bismuth trichloride
89485-06
89485-14
CAAA89485-06EA
112.81
CAD
CAAA89485-06
CAAA89485-14
Bismuth(III) chloride, anhydrous ≥99.999% (metals basis)
Bismuth (III) chloride
MDL: MFCD00003461
Fieser: 7,24 15,37 18,52 19,37 20,45 21,59
Purity: 99.999%
CAS No.: 7787-60-2
Molecular Formula: BiCl3
Formula Weight: 315.34
Physical Form: Crystalline
Density: 4.75
Appearance: White
Melting Point: 230-232°
Boiling Point: ca 430° subl.
Merck Reference: 14,1262
UN No.: 3260
MDL No.: MFCD00003461
Formula:
BiCl₃ MW: 315.34 g/mol Boiling Pt: 447 (subl.) Melting Pt: 232 Density: 4.75 Storage Temperature: Ambient |
MDL Number:
MFCD00003461 CAS Number: 7787-60-2 EINECS: 232-123-2 UN: 3260 ADR: 8 III Merck Index: 14,1262 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...