Rhenium disulfide 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Rhenium (IV) sulphide, Rhenium (IV) sulfide
89482-04
89482-09
CAAA89482-04EA
635.16
CAD
CAAA89482-04
CAAA89482-09
Rhenium disulfide 99%
Rhenium disulphide
MDL: MFCD00016307
Purity: 99%
CAS No.: 12038-63-0
Molecular Formula: ReS2
Formula Weight: 250.33
Physical Form: Powder
Density: 7.506
MDL No.: MFCD00016307
Formula:
ReS₂ MW: 250.34 g/mol Density: 7.506 Storage Temperature: Ambient |
MDL Number:
MFCD00016307 CAS Number: 12038-63-0 EINECS: 234-878-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...