Silver (I) sulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

89473-06 89473-14
CAAA89473-06EA 118.82 CAD
CAAA89473-06 CAAA89473-14
Silver (I) sulfide ≥99.9% (metals basis)
Silver (I) sulphide

MDL: MFCD00003406


Purity: 99.9+% CAS No.: 21548-73-2 Molecular Formula: Ag2S Formula Weight: 247.80 Physical Form: Crystalline powder Density: 7.317 Appearance: Dark grey to black Odor: Odorless Melting Point: 825° Merck Reference: 14,8530 UN No.: 3077 MDL No.: MFCD00003406

Formula: Ag₂S
Melting Pt: 825 °C
Density: 7.317
Storage Temperature: Ambient
MDL Number: MFCD00003406
CAS Number: 21548-73-2
EINECS: 244-438-2
UN: 3077
ADR: 9,III
Merck Index: 14,8530

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR