Silicon(II) oxide ≥99.8% (metals basis), Optical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
89430-18
89430-30
CAAA89430-18EA
180.12
CAD
CAAA89430-18
CAAA89430-30
Silicon(II) oxide ≥99.8% (metals basis), Optical Grade
Silicon (II) oxide
MDL: MFCD00151536
Purity: 99.8%
CAS No.: 10097-28-6
Molecular Formula: OSi
Formula Weight: 44.09
Physical Form: -325 Mesh Powder (typically 10 micron or less)
Density: 2.13
Appearance: Black
Odor: Odorless
Melting Point:
Formula:
SiO Boiling Pt: 1880 °C Melting Pt: >1702 °C Density: 2.13 Storage Temperature: Ambient |
MDL Number:
MFCD00151536 CAS Number: 10097-28-6 EINECS: 233-232-8 Merck Index: 14,8496 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...