Sulfuryl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sulphuryl dichloride, Sulfuryl dichloride

89411-A1 89411-30
CAAAA89411-A1EA 873.74 CAD
CAAAA89411-A1 CAAA89411-30
Sulfuryl chloride 97%
Sulphuryl chloride

Chlorinating, sulfonating, and chlorosulfonating agent


Purity: 97% CAS No.: 7791-25-5 Molecular Formula: Cl2O2S Formula Weight: 134.96 Physical Form: Liquid Density: 1.680 Refractive Index: 1.4437 Sensitivity: Moisture Sensitive Appearance: Colorless to pale yellow Odor: Pungent Melting Point: -54° Boiling Point: 69° Solubility: Decomposes in water. Miscible with benzene, toluene, ether, glacial acetic acid Merck Reference: 14,8980 UN No.: 1834 MDL No.: MFCD00011451


Chlorinating, sulfonating, and chlorosulfonating agent

Formula: Cl₂O₂S
MW: 134.97 g/mol
Boiling Pt: 69.1
Melting Pt: -54
Density: 1.680
Storage Temperature: Ambient
MDL Number: MFCD00011451
CAS Number: 7791-25-5
EINECS: 232-245-6
UN: 1834
ADR: 6.1 I
Merck Index: 14,8980

Order Now

Specification Test Results

Form Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR