Kieselguhr -325 Mesh Powder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Siliceous earthDiatomiteInfusorial earthDiatomaceous silicaCeliteDiatomaceous earth

89381-A9 89381-A9 89381-A1 89381-A1
CAAA89381-A9EA 562.48 CAD
AA89381-A9 CAAA89381-A9 AA89381-A1 CAAA89381-A1
Kieselguhr -325 Mesh Powder
Kieselguhr

Kieselguhr is used in used as a filtration aid, mild abrasive in products including metal polishes and toothpaste, absorbent for liquids, matting agent for coatings, reinforcing filler in plastics and rubber, anti-block in plastic films, porous support for chemical catalysts, cat litter, activator in blood clotting studies, a stabilizing component of dynamite, and a thermal insulator.

Formula: O₂Si
MW: 60.08 g/mol
Melting Pt: >1300 °C (dec.)
Density: 2.0
Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 7631-86-9
EINECS: 231-545-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR