Silicon(IV) oxide, amorphous ≥99.8%, fumed S.A. 350-420m2/g
Supplier: Thermo Scientific Chemicals
|
Warning
|
89376-18
89376-30
89376-A1
CAAA89376-18EA
35.71
CAD
CAAA89376-18
CAAA89376-30
CAAA89376-A1
Silicon(IV) oxide, amorphous ≥99.8%, fumed S.A. 350-420m2/g
Silica gel amorphous
MDL: MFCD00011232
CAS No.: 7631-86-9
Molecular Formula: SiO2
Formula Weight: 60.09
Physical Form: -325 Mesh Powder
Density: 2.2
Refractive Index: 1.46
Odor: Odorless
Melting Point: 1710°
Boiling Point: 2230°
Merck Reference: 14,8493
MDL No.: MFCD00011232
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: 1710 °C Density: 2.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 231-545-4 Merck Index: 14,8493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...