Ammonium sulfate ≥99.95% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

89363-22 89363-36 89363-A3
CAAA89363-22EA 89.59 CAD
CAAA89363-22 CAAA89363-36 CAAA89363-A3
Ammonium sulfate ≥99.95% (metals basis)
Ammonium sulphate

MDL: MFCD00003391


Purity: 99.95% CAS No.: 7783-20-2 Molecular Formula: H8N2O4S Formula Weight: 132.14 Physical Form: Crystalline Density: 1.769 Appearance: White Melting Point: ca 280° dec. Merck Reference: 14,555 MDL No.: MFCD00003391

Formula: (NH₄)₂SO₄
Melting Pt: ∼ 280 °C (dec.)
Density: 1.769
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 14,555

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR