Cesium chloride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
|
89188-09
89188-18
CAAA89188-09EA
161.16
CAD
CAAA89188-09
CAAA89188-18
Cesium chloride ≥99.999% (metals basis)
Cesium chloride
MDL: MFCD00010955
Purity: 99.999+%
CAS No.: 7647-17-8
Molecular Formula: CsCl
Formula Weight: 168.36
Physical Form: Powder
Density: 3.99
Refractive Index: 1.6418
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 646°
Boiling Point: 1303°
Merck Reference: 14,2011
MDL No.: MFCD00010955
Formula:
CsCl Boiling Pt: 1303 °C Melting Pt: 646 °C Density: 3.99 Storage Temperature: Ambient |
MDL Number:
MFCD00010955 CAS Number: 7647-17-8 EINECS: 231-600-2 Merck Index: 14,2011 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...