Tin(II) telluride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
89186-09
89186-18
CAAA89186-09EA
194.34
CAD
CAAA89186-09
CAAA89186-18
Tin(II) telluride ≥99.999% (metals basis)
Tin (II) telluride
MDL: MFCD00049543
Purity: 99.999%
CAS No.: 12040-02-7
Molecular Formula: SnTe
Formula Weight: 246.29
Physical Form: 3-12mm Pieces
Density: 6.48
Melting Point: 780°
UN No.: 3288
MDL No.: MFCD00049543
Formula:
SnTe Melting Pt: 780 °C Density: 6.48 Storage Temperature: Ambient |
MDL Number:
MFCD00049543 CAS Number: 12040-02-7 EINECS: 234-914-8 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...