Titanium(II) hydride ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
For preparation of titanium compounds and coatings, in formation of glass and ceramic seals, as a catalyst, for thermal storage, as a source for pure H2
Purity: 99%
CAS No.: 7704-98-5
Molecular Formula: H2Ti
Formula Weight: 49.92
Physical Form: 1-3 Micron Powder
Density: 3.9
Appearance: Dark grey
Odor: Odorless
Melting Point: 450° dec.
Merck Reference: 14,9473
UN No.: 1871
MDL No.: MFCD00151301
For preparation of titanium compounds and coatings, in formation of glass and ceramic seals, as a catalyst, for thermal storage, as a source for pure H2
Formula:
H₂Ti Melting Pt: 450 °C (dec.) Density: 3.9 Storage Temperature: Ambient |
MDL Number:
MFCD00151301 CAS Number: 7704-98-5 EINECS: 231-726-8 UN: 1871 ADR: 4.1,II Merck Index: 14,9473 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...