Sodium tetraethylborate 98%

Supplier: Thermo Scientific Chemicals
Danger

89137-03 89137-06
CAAA89137-03EA 263.86 CAD
CAAA89137-03 CAAA89137-06
Sodium tetraethylborate 98%
Sodium tetraethylborate

MDL: MFCD00061547 Dissolves in water with decomposition


Purity: 98% CAS No.: 15523-24-7 Molecular Formula: C8H20BNa Formula Weight: 150.05 Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Appearance: White Melting Point: dec. Solubility: Dissolves in water with decomposition UN No.: 3393 MDL No.: MFCD00061547

Formula: C₈H₂₀BNa
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00061547
CAS Number: 15523-24-7
UN: 3393
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR