Aluminium bromide ≥99.997% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum bromide
89065-06
89065-14
CAAA89065-06EA
90.38
CAD
CAAA89065-06
CAAA89065-14
Aluminium bromide ≥99.997% (metals basis)
Aluminium bromide
MDL: MFCD00003421
Fieser: 1,22 2,19 3,7 4,10 5,10 6,17 9,11
Purity: 99.997%
CAS No.: 7727-15-3
Molecular Formula: AlBr3
Formula Weight: 266.71
Physical Form: Crystalline
Density: 3.21
Appearance: White to orange
Odor: Pungent
Melting Point: 97.5°
Boiling Point: 265°
Merck Reference: 14,332
UN No.: 1725
MDL No.: MFCD00003421
Formula:
AlBr₃ MW: 266.69 g/mol Boiling Pt: 265 Melting Pt: 97 Density: 3.21 Storage Temperature: Ambient |
MDL Number:
MFCD00003421 CAS Number: 7727-15-3 EINECS: 231-779-7 UN: 1725 ADR: 8 II Merck Index: 14,332 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...