Sodium trimetaphosphate

Supplier: Thermo Scientific Chemicals

89063-A1 89063-A7
CAAA89063-A1EA 107.28 CAD
CAAA89063-A1 CAAA89063-A7
Sodium trimetaphosphate
Trisodium trimetaphosphate

MDL: MFCD00867826


CAS No.: 7785-84-4 Molecular Formula: (NaPO3)3 Formula Weight: 305.89 Physical Form: Powder Density: 2.49

Formula: Na₃O₉P₃
Melting Pt: 588 °C
Density: 2.49
Storage Temperature: Ambient
MDL Number: MFCD00867826
CAS Number: 7785-84-4
EINECS: 232-088-3
Merck Index: 14,8696

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR