Potassium hexafluorosilicate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Potassium fluorosilicate
89023-A1
89023-A7
CAAA89023-A1EA
127.98
CAD
CAAA89023-A1
CAAA89023-A7
Potassium hexafluorosilicate 98%
Potassium hexafluorosilicate
MDL: MFCD00040666
Slightly soluble in cold water. Hydrolyzes in hot water. Insoluble in alcohol.
Purity: 98%
CAS No.: 16871-90-2
Molecular Formula: K2SiF6
Formula Weight: 220.28
Physical Form: Powder
Density: 2.27
pH: 3.4
Melting Point: dec.
Solubility: Slightly soluble in cold water. Hydrolyzes in hot water. Insoluble in alcohol
Merck Reference: 14,7638
UN No.: 2655
MDL No.: MFCD00040666
Formula:
F₆K₂Si MW: 220.27 g/mol Melting Pt: Dec. Density: 2.27 Storage Temperature: Ambient |
MDL Number:
MFCD00040666 CAS Number: 16871-90-2 EINECS: 240-896-2 UN: 2655 ADR: 6.1 III Merck Index: 14,7638 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...