Hydroxylammonium sulfate ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
88944-36
88944-A3
CAAA88944-36EA
93.22
CAD
CAAA88944-36
CAAA88944-A3
Hydroxylammonium sulfate ≥99%
Hydroxylammonium sulphate
MDL: MFCD00044869
Soluble in water
Purity: 99%
CAS No.: 10039-54-0
Molecular Formula: H3NO.0.5H2SO4
Formula Weight: 164.14
Physical Form: Crystalline
Density: 1.88
Appearance: White
Odor: Odorless
pH: 3.6
Melting Point: ca 170° dec.
Solubility: Soluble in water
Merck Reference: 14,4828
UN No.: 2865
MDL No.: MFCD00044869
Formula:
H₈N₂O₆S Melting Pt: ∼ 170 °C (dec.) Density: 1.88 Storage Temperature: Ambient |
MDL Number:
MFCD00044869 CAS Number: 10039-54-0 EINECS: 233-118-8 UN: 2865 ADR: 8,III Merck Index: 14,4828 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...