Charcoal activated, slug 6-8 mesh
Supplier: Thermo Scientific Chemicals
Synonyms:
Carbon activated, Activated carbon, Activated charcoal
88764-A1
88764-A7
CAAA88764-A1EA
126.54
CAD
CAAA88764-A1
CAAA88764-A7
Charcoal activated, slug 6-8 mesh
Charcoal activated
For hydrocarbon recovery
MDL No.: MFCD00133992
For hydrocarbon recovery
Formula:
C MW: 12.01 g/mol Melting Pt: 3550 Density: 2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-153-3 UN: 1362 ADR: 4.2 III REACH: 01-2119488894-16 Merck Index: 13,01818 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...