Silver trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
88722-04
88722-09
88722-18
CAAA88722-04EA
45.35
CAD
CAAA88722-04
CAAA88722-09
CAAA88722-18
Silver trifluoromethanesulfonate 98%
Silver trifluoromethanesulphonate
MDL: MFCD00013226
Purity: 98%
CAS No.: 2923-28-6
Molecular Formula: CAgF3O3S
Formula Weight: 256.93
Physical Form: Crystalline
Sensitivity: Light Sensitive
Appearance: White to grey
Odor: Odorless
Melting Point: 356°
UN No.: 3260
MDL No.: MFCD00013226
Formula:
CAgF₃O₃S Melting Pt: 356 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013226 CAS Number: 2923-28-6 EINECS: 220-882-2 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...