Sodium thiosulfate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution

Supplier: Thermo Scientific Chemicals

88628-LQ 88628-LS
CAAA88628-LQEA 102.7 CAD
CAAA88628-LQ CAAA88628-LS
Sodium thiosulfate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Sodium thiosulphate

MDL: MFCD00003499 Notes: One unit makes one liter of 0.1N solution after dilution


CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Physical Form: Liquid MDL No.: MFCD00003499

Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR