Disodium oxalate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances

88626-LQ 88626-LS
CAAA88626-LQEA 121.66 CAD
CAAA88626-LQ CAAA88626-LS
Disodium oxalate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
di-Sodium oxalate

MDL: MFCD00012465 Notes: One unit makes one liter of 0.1N solution after dilution


CAS No.: 62-76-0 Molecular Formula: Na2C2O4 Formula Weight: 134.00 Physical Form: Liquid Odor: Odorless MDL No.: MFCD00012465

Formula: Na₂C₂O₄
MW: 134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Merck Index: 13,08723

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR