Silver nitrate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
88614-LQ
88614-LS
CAAA88614-LQEA
194.34
CAD
CAAA88614-LQ
CAAA88614-LS
Silver nitrate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Silver nitrate
MDL: MFCD00003414
Notes: One unit makes one liter of 0.1N solution after dilution
CAS No.: 7761-88-8
Molecular Formula: AgNO3
Formula Weight: 169.87
Physical Form: Liquid
Sensitivity: Light Sensitive
Appearance: Colorless
UN No.: 3093
MDL No.: MFCD00003414
Formula:
AgNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 3093 ADR: 8,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...