Potassium hydroxide concentrate for dilution to 0.5 N in aqueous solution, Acculute™ standard volumetric solution

Supplier: Thermo Scientific Chemicals
Danger

88602-LQ 88602-LS
CAAA88602-LQEA 118.03 CAD
CAAA88602-LQ CAAA88602-LS
Potassium hydroxide concentrate for dilution to 0.5 N in aqueous solution, Acculute™ standard volumetric solution
Potassium hydroxide

MDL: MFCD00003553 Notes: One unit makes one liter of 0.5N solution after dilution


CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Physical Form: Liquid Appearance: Colorless UN No.: 1814 MDL No.: MFCD00003553

Formula: KOH
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR