Indium antimonide ≥99.99% (metals basis), Electronic Grade for the electronics industry
Supplier: Thermo Scientific Chemicals
|
Warning
|
88460-04
88460-09
88460-18
CAAA88460-04EA
69.2
CAD
CAAA88460-04
CAAA88460-09
CAAA88460-18
Indium antimonide ≥99.99% (metals basis), Electronic Grade for the electronics industry
Indium antimonide
In semiconductor electronics, as an infrared detector and filter
Purity: 99.99%
CAS No.: 1312-41-0
Molecular Formula: InSb
Formula Weight: 236.57
Physical Form: Pieces
Density: 5.75 at m.p.
Melting Point: 535°
Merck Reference: 14,4948
UN No.: 1549
MDL No.: MFCD00016146
In semiconductor electronics, as an infrared detector and filter
Formula:
InSb MW: 236.58 Melting Pt: 527 Density: 5.75 (m.p. Storage Temperature: Ambient |
MDL Number:
MFCD00016146 CAS Number: 1312-41-0 EINECS: 215-192-3 UN: 1549 ADR: 6.1 III Merck Index: 14,4948 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...