Gallium arsenide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

88458-04 88458-09 88458-18
CAAA88458-04EA 195.92 CAD
CAAA88458-04 CAAA88458-09 CAAA88458-18
Gallium arsenide ≥99.999% (metals basis)
Gallium arsenide

Semiconductors (transistors, lasers, solar cells)


Purity: 99.999% CAS No.: 1303-00-0 Molecular Formula: GaAs Formula Weight: 144.64 Physical Form: 25mm & Down Polycrystalline Pieces Density: 5.31 Melting Point: 1238° Merck Reference: 14,4347 UN No.: 1557 MDL No.: MFCD00011017


Semiconductors (transistors, lasers, solar cells)

Formula: AsGa
MW: 144.64
Melting Pt: 1238
Density: 5.31
Storage Temperature: Ambient
MDL Number: MFCD00011017
CAS Number: 1303-00-0
EINECS: 215-114-8
UN: 1557
ADR: 6.1 II
Merck Index: 14,4347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR